Learn More
ABclonal Technology TOMM34 Rabbit mAb
SDP
Supplier:  ABclonal Technology A27418100UL
Encompass_Preferred
The protein encoded by this gene is involved in the import of precursor proteins into mitochondria The encoded protein has a chaperone-like activity binding the mature portion of unfolded proteins and aiding their import into mitochondria This protein which is found in the cytoplasm and sometimes associated with the outer mitochondrial membrane has a weak ATPase activity and contains 6 TPR repeats
Catalog No. 50-000-09696