Learn More
Thermo Scientific Gallery System Reagents for Environmental and Industrial Analysis | | 1 EA
Supplier:  Thermo Scientific 984372
Encompass
Save time by eliminating reagent preparation; Over 40 ready-to-use gallery system reagents; Automated dilutions for a wide range of analyte concentrations; Based on standard water and wastewater methods
Catalog No. NC2232266