Learn More
Apexbio Technology LLC Tonabersat 175013-84-0 50mg
SDP
Supplier:  Apexbio Technology LLC A388350
Encompass_Preferred
Tonabersat (CAS 175013-84-0) is a gap-junction modulator targeting neuronal and satellite glial cell communication within the trigeminal ganglion It is designed to modulate gap-junction pathways thereby regulating neurological signaling associated with migraine aura and epilepsy Tonabersat exerts its biological activity primarily through inhibition of gap-junction communication leading to decreased cortical spreading depression (CSD) a process implicated in migraine pathophysiology Based on these pharmacological properties Tonabersat holds research potential in the study of migraine and epilepsy
Catalog No. 50-000-96799