Learn More
Med Vet International TonoVet Tonometer, J1000, Each
Supplier:  Med Vet International JOR1000
Encompass_Preferred
TonoVet The use of tonometry to measure Intra Ocular Pressure (IOP) is the only real tool for the screening of glaucoma in veterinary patients. The TonoVet has been used on all veterinary patients including equine. The TonoVet is self-calibrating with its own software. Why the TonoVet? - It is easy to use and fully automated - Faster results - More comfortable for the patient, no topical anesthetic - Higher accuracy at higher and lower IOP - Longer warranty - 2 years - In clinic training available - Held upright in a natural, ergonomic position like an - Instructional DVD included opthalmoscope versus horizontally like a flashlight. Specifications Weight: 155g (5oz) without batteries; 250g (8oz) with batteries Power Supply: 4 x AA batteries (approx. 1,000 measurements per battery set) Measurement Range: 1-99 mm Hg Size: 13-32mm W x 45-80mm D x 230mm L Padded Storage Case Provided Two year limited warranty Extra probes (100 per package)
Catalog No. 50-004-95935