Learn More
Electron Microscopy Sciences Micro-Tool Micro-Spade, 0.025 mm, Tip 3
SDP
Supplier:  Electron Microscopy Sciences 62091040.025
Encompass
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.025 mm
Tips: Tip 3
Catalog No. 50-996-607