Learn More
Tecan DiTi Transfer Tool
Supplier:  Tecan 30083403
Encompass_Preferred
Nested Liquid Handling (LiHa) Disposable Tip (DiTi) Transfer tool makes it possible to benefit from the space-saving advantages of nested LiHa disposable tips without requiring a gripper to remove empty tip trays. This patent pending solution is based on the pick-up and disposal of empty tip trays using a unique consumable the Disposable Transfer Tool and is compatible with the Tecan Freedom EVO Liquid Handling Arm, the Fluent Flexible Channel Arm™ (FCA). The use of these tools requires the Transfer Tool Holder p/n 30092506 which fits into a three-position trough carrier. 112 pieces
Catalog No. NC0775975