Learn More
Electron Microscopy Sciences Tooled Neck Vials, 2 Dram
SDP
Supplier:  Electron Microscopy Sciences 7264602
Encompass_Preferred
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with an open-bottom closure.
Volume: 2 Dram
Catalog No. 50-949-471