Learn More
Electron Microscopy Sciences Tooled Neck Vials, 3 Dram
SDP
Supplier:  Electron Microscopy Sciences 7264603
Encompass_Preferred
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with a closed bottom, two-piece closure.
Volume: 3 Dram
Catalog No. 50-949-507