Learn More
Medchemexpress LLC Toosendanin | 58812-37-6 | 98.7% | 574.62 | C30H38O11 | 10 MG

Supplier: Medchemexpress LLC HYN026310MG
Toosendanin is a triterpenoid natural product extracted from the bark or fruit of Melia toosendan Sieb. et Zucc., used in research for its analgesic, insecticidal, and anti-inflammatory activities.
- Natural triterpenoid isolated from Melia toosendan.
- Exhibits analgesic, insecticidal, and anti-inflammatory activity.
- High purity: 98.74%.
- Molecular weight: 574.62.
- Appearance: solid, white to off-white.
- Soluble in DMSO (100 mg/mL; may require ultrasonic), with demonstrated in vivo formulation in 10% DMSO/90% corn oil.
- Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month), protect from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.