Learn More
Med Vet International Tooth Punch-Curved, 20cm, J0713
Supplier: Med Vet International JOR713
The punch is used for removal of cheek teeth through a trephined hole into the sinus. A knurled handle gives a better grip. A round head with a slight recessed area fits better onto the tooth. Stainless steel. 20cm L.