Learn More
Cayman Chemical TopIramate-d12 500ug
SDP
Supplier:  Cayman Chemical 26443500 UG
Encompass_Preferred
An internal standard for the quantification of topiramate by GC- or LC-MS
Catalog No. 50-293-5276