Learn More
Selleck Chemical LLC Topiramate 100mg 97240-79-4 MCN 4853, RWJ 17021
SDP
Supplier:  Selleck Chemical LLC S1438100MG
Encompass_Preferred
Topiramate (MCN 4853, RWJ 17021) is a mutil-targeted inhibitor, including voltage-gated sodium channel and calcium channel, AMPA/kainate receptor and carbonic anhydrase, used to treat epilepsy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-767-8