Learn More
Medchemexpress LLC Topoisomerase II inhibitor 13 | 451515-89-2 | C22H23N9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1534015MG
Encompass_Preferred
Topoisomerase II inhibitor 13 is a topoisomerase II (Topo II) inhibitor. It demonstrates antiproliferative activity against various cancer cells and induces apoptosis in cancer cells.
  • Topoisomerase II (Topo II) inhibitor.
  • Shows antiproliferative activity against several cancer cells.
  • Induces cancer cell apoptosis.
  • Exerts strong inhibitory effect on Topo II activity at 20 μM, and almost completely inhibits Topo II activity at 50 μM.
  • Inhibits cancer cell proliferation with varying IC50s across different cell lines (e.g., A549, K562, Raji, HL-60, MDA-MB-231).
Catalog No. 50-003-73269