Learn More
Cayman Chemical TopotecnhydrochlorIde 50mg
SDP
Supplier:  Cayman Chemical 1412950 MG
Encompass_Preferred
An inhibitor of DNA topoisomerase I and a derivative of camptothecin; inhibits DNA topoisomerase I in human MCF-7 breast and DU-145 prostate cancer cells (IC50s = 13 and 2 nM, respectively); induces cytotoxicity and DNA damage in HT-29 human colon adenocarcinoma cells (IC50s = 33 and 280 nM, respectively)
Catalog No. 50-295-3579