Learn More
Medchemexpress LLC Topovale (ARC-111) | 500214-53-9 | 99.9% | 421.45 | C23H23N3O5 | 50 MG

Supplier: Medchemexpress LLC HY11894250MG
Topovale (ARC-111) is a potent topoisomerase I inhibitor used in preclinical research to induce reversible TOP1 cleavage complexes and cause low nanomolar cytotoxicity in cancer cell models.
- Potent topoisomerase I inhibitor with low nanomolar cytotoxicity.
- High reported purity (99.94%).
- Molecular formula C23H23N3O5; molecular weight 421.45.
- Soluble in DMSO: 4.03 mg/mL (adjust pH to 2 with TFA; ultrasonic).
- Powder stable at -20°C for up to 3 years; limited stability in solvent.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.