Learn More
Medchemexpress LLC Topramezone | 210631-68-8 | 100.0% | 363.39 g/mol | C16H17N3O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1449005MG
Encompass_Preferred
Topramezone (Standard) is an analytical reference standard of topramezone, a 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor and post-emergence herbicide used for control of broadleaf and grass weeds in corn. Supplied for research and analytical applications, it is intended for qualitative and quantitative methods in HPLC, GC, and MS. CAS 210631-68-8; formula C16H17N3O5S; molecular weight 363.39 g/mol.
  • Intended for analytical and research use in HPLC, GC, and MS.
  • Potent 4-HPPD inhibitor used as a herbicide reference standard.
  • Characterized by CAS 210631-68-8 and defined molecular formula and weight.
  • Provided as an analytical-grade standard with high purity for quantitative assays.
  • Suitable as a reference material for method development and validation.
Catalog No. 50-002-21195