Learn More
Glass Expansion Torch Adaptor for Ball Joint Injector. Cassette Mount Adaptor for Ball Joint Injector. This adaptor is suitable for injectors with a ball joint. Compatible with PerkinElmer Elan DRC versions, NexION 300/350.
SDP
Supplier:  Glass Expansion 318082824
Encompass_Preferred
Cassette Mount Adaptor for Ball Joint Injector (W1012406). This adaptor is suitable for injectors with a ball joint. This torch adaptor is usually used with PerkinElmer Elan DRC models.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3154