Learn More
Selleck Chemical LLC Torin 1 10mg 1222998-36-8
SDP
Supplier:  Selleck Chemical LLC S282710MG
Encompass_Preferred
Torin 1 is a potent inhibitor of mTORC1/2 with IC50 of 2 nM/10 nM in cell-free assays; exhibits 1000-fold selectivity for mTOR than PI3K. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3036