Learn More
Cayman Chemical Torin 1, 1222998-36-8, 10 mg
SDP
Supplier:  Cayman Chemical 1099710
Encompass_Preferred
Molecular Formula C35H28F3N5O2, Purity ≥95%, Formula Weight 607.6; A potent and selective inhibitor of mTOR (IC50 = 2 and 10 nM for mTORC1 and mTORC2, respectively); efficacious at a dose of 20 mg/kg in inhibiting downstream effectors of mTOR in mice.
Catalog No. NC0418592