Learn More
Medchemexpress LLC HY-13002 10mg Medchemexpress, Torin 2 CAS:1223001-51-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1300210MG
Encompass_Preferred
Medchemexpress, HY-13002 10mg Torin 2 CAS:1223001-51-1 Torin 2 is an mTOR inhibitor with EC50 of 0.25 nM for inhibiting cellular mTOR activity, and exhibits 800-fold selectivity over PI3K (EC50: 200 nM). Torin 2 also inhibits DNA-PK with an IC50 of 0.5 nM in the cell free assay. Torin 2 can suppress both mTORC1 and mTORC2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8934