Learn More
Medchemexpress LLC Torin 2 | 1223001-51-1 | 99.65% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1300210MM/1ML
Encompass_Preferred
Torin 2 is an mTOR inhibitor with an EC50 of 0.25 nM for inhibiting cellular mTOR activity. It exhibits 800-fold selectivity over PI3K (EC50: 200 nM) and also inhibits DNA-PK with an IC50 of 0.5 nM in the cell-free assay. Torin 2 can suppress both mTORC1 and mTORC2.
  • Potent mTOR inhibitor with an EC50 of 0.25 nM for cellular mTOR activity.
  • Exhibits 800-fold selectivity over PI3K.
  • Inhibits DNA-PK with an IC50 of 0.5 nM in cell-free assays.
  • Suppresses both mTORC1 and mTORC2.
  • Demonstrates good bioavailability and exposure in vivo.
  • Possesses improved pharmacokinetic properties for in vivo experiments.
  • Maintains strong inhibition of mTOR activity in lung and liver for extended periods.
Catalog No. 50-003-38215