Learn More
Medchemexpress LLC Toripalimab 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP997810MG
Encompass_Preferred
Toripalimab is a selective recombinant humanized monoclonal antibody against PD-1 Toripalimab is able to bind to PD-1 and block the interaction with its ligands Toripalimab has exhibited primary anti-tumor effects in tumors such as melanoma lung cancer digestive tract tumors hepatobiliary and pancreatic tumors neuroendocrine neoplasms nasopharyngeal carcinoma and urothelial carcinoma[1]
Catalog No. 50-003-22307