Learn More
Enterprise Technology Solutions TORK Air Freshener Dispenser,Elevation Design,Propellant-Free,Scented Oils,Non-Flammable,6/CT,White
SDP
Supplier:  Enterprise Technology Solutions TRK256021
Encompass_Preferred
Tork Constant Air Freshener Dispenser in Elevation Design uses a unique, high-tech distribution technology to eliminate bad odors. It's certified "easy-to-use" for effortless refilling. This propellant-free system uses only scented oils, and it's not pressurized or classified as flammable. Available with three carefully selected scents (not included) - Breeze, Blossom and Odor Neutralizer - for constant freshness.
Catalog No. 50-007-90228