Learn More
Medchemexpress LLC Torsemide-d7 | 1189375-06-1 | >99.7% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0247S5MG
Encompass_Preferred
Torsemide-d7 is the deuterium-labeled form of Torsemide, an orally active loop diuretic. It exhibits anti-aldosterone and vasodilatory effects, making it suitable for research into heart failure, renal disease, and hepatic cirrhosis. Deuteration incorporates stable heavy isotopes into drug molecules, primarily as tracers for quantitation during drug development and to study pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Exhibits anti-aldosterone effects
  • Possesses vasodilatory effects
  • Applied in research for heart failure
  • Utilized in renal disease research
  • Investigated for hepatic cirrhosis studies
Catalog No. 50-004-07393