Learn More
Waters Corp Waters Column, 1-AA, 1.7 μm, 3 mm X 50 mm, 130 Å
Supplier:  Waters Corp 186007627
Encompass_Preferred
Torus Columns are specifically designed to use the complete range of capabilities of the ACQUITY UPC2 System to achieve fast, robust achiral separations. These columns simplify the method development process with four completely new and innovative 1.7 μm chemistries for convergence chromatography. They are designed for excellent peak shape that eliminates or reduces the need for additives and offer added selectivity for a wide range of compounds and improved robustness.
  • Designed for use with the ACQUITY UPC2 system.
  • Utilizes four new and innovative 1.7 μm chemistries for convergence chromatography.
  • Simplifies method development.
  • Provides excellent peak shape, potentially reducing the need for additives.
  • Offers added selectivity for a wide range of compounds.
  • Improved robustness.
Catalog No. 50-203-8126