Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp OBD Prep Column, 2-PIC, 130 Å, 5 μm

Supplier:  Waters Corp 186008597

Encompass_Preferred

This preparative column is designed for general use in supercritical fluid chromatography (SFC) separations, especially for acidic and basic compounds. It offers enhanced performance, including improved peak shape, increased retention, and novel selectivity. It also provides superior robustness for laboratory-scale purification and supports method scale-up.

  • Spherical particle shape
  • Particle sizes of 1.7 μm and 5 μm available
  • Pore volume of 0.7 cc/g
  • Pore size of 130 Å
  • Surface area of 185 m²/g
  • 2-picolylamine chemistry
  • Designed for general use with acidic and basic compounds
  • Improved peak shape
  • Increased retention
  • Novel selectivity
  • Enhanced performance over conventional phases
  • Suitable for laboratory-scale purification
  • Supports method scale-up

Catalog No. 50-239-2440


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.