Learn More
Waters Corp Preparative Column, 2-Picolylamine (2-PIC), 5 μm, 19 × 150 mm
Supplier:  Waters Corp 186008587
Encompass_Preferred
This OBD 19 × 150 mm Torus 5 μm Preparative Column, featuring 2-PIC chemistry, is designed for general use in separating acidic and basic compounds.
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles.
  • Hydrophilic surface for controlled retention and minimized unwanted interactions.
  • Provides individual selectivity and improved peak shape for acidic and basic compounds.
  • Offers novel selectivity versus conventional 2-ethylpyridine (2-EP) phases.
  • Enhanced retention for diverse applications.
  • Suitable for method transfer and scale-up from analytical to preparative SFC.
  • Robust for laboratory-scale purification, maintaining retention and selectivity over time.
  • Patented OBD Technology contributes to excellent column lifetimes.
Catalog No. 50-239-2617