Learn More
Waters Corp Prep Column, 2-PIC, 5 μm, 19 mm, 100 mm
Supplier:  Waters Corp 186008586
Encompass_Preferred
Torus 2-PIC OBD Prep Columns are designed to streamline SuperCritical Fluid Chromatography (SFC) from analytical to preparative scales. These columns offer a wide range of selectivity and are available in various formats for SFC applications.
  • 2-PIC chemistry
  • SuperCritical Fluid (SFC) separation mode
  • Hybrid particle substrate
  • Spherical particle shape
  • OBD prep column format
  • SFC preparative system
  • SFC and SFC/MS techniques
  • 1 unit per package
  • 130 Å pore size
  • 5 μm particle size
  • 19 mm inner diameter
  • 100 mm length
  • 60 °C temperature limit
  • 6000 psi (415 Bar) maximum pressure
  • 185 surface area
  • Not endcapped
  • Endfitting type: Waters
Catalog No. 50-239-1371