Learn More
Waters Corp Waters Corporation Column, Torus 2-PIC, 130Å, 1.7 μm, 3 mm × 100 mm
Supplier:  Waters Corp 186007602
Encompass_Preferred
Torus Columns are designed for fast, robust achiral separations with ACQUITY UPC2 System. They simplify method development using four innovative 1.7 μm chemistries for convergence chromatography. These columns provide excellent peak shape, reduce additive needs, and offer enhanced selectivity and improved robustness. The Torus 2-PIC Column is ideal for acidic and basic compounds, and can analyze acid degradation products of formulated omeprazole tablets.
  • Packing materials compatible with ACQUITY UPC2 and SFC purification systems.
  • Manufactured using ultra-pure reagents in an ISO-certified facility.
  • Batches examined for repeatable performance; results held to limited specification ranges.
  • Performance Chromatogram and Batch Analysis Certificate issued for each column.
  • eCord™ Intelligent Chip provides product information for 1.7 μm Columns.
Catalog No. 50-222-4191