Learn More
Waters Corp Torus Preparative Column, 5 μm, 50 x 50 mm, DEA Chemistry
Supplier:  Waters Corp 186008628
Encompass_Preferred
The Torus preparative column combines state-of-the-art media manufacturing with industry-leading column technology to provide robust laboratory-scale purification. Designed for achiral SFC separations, it offers a wide range of selectivity and excellent peak shape, making it suitable for method transfer and scale-up.
  • Provides complementary selectivity and superior peak shape for very strong bases
  • Suitable for achiral SFC separations
  • Offers a wide range of selectivity and excellent peak shape
  • Suited for method transfer and scale-up
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles
  • Imparts a new level of robustness to laboratory-scale purification
Catalog No. 50-239-2582
Add to Cart
Request a Quote