Learn More
Waters Corp Waters Torus Diol (OH) Column, 130 Å, 1.7 μm, 3 mm x 150 mm
Supplier:  Waters Corp 186007612
Encompass_Preferred
Torus Columns are specifically designed to leverage the full capabilities of the ACQUITY UPC2 System for fast, robust achiral separations. These columns simplify method development through innovative 1.7 μm chemistries for convergence chromatography. They deliver excellent peak shape, often eliminating or reducing the need for additives, and provide enhanced selectivity for a broad range of compounds, coupled with improved robustness.
  • Designed for fast, robust achiral separations with excellent peak shape and high selectivity.
  • Simplifies method development with innovative 1.7 μm chemistries for convergence chromatography.
  • High-density diol surface bonding provides enhanced durability and consistent performance.
  • Features patent-pending two-stage functionalization of ethylene bridging hybrid (BEH) particles.
  • Optimized for quality control laboratories, ensuring reproducible results.
Catalog No. 50-203-8112