Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Torus OBD Preparative Column, DIOL, 130 Å, 5 μm

Supplier:  Waters Corp 186008603

Encompass_Preferred

The Torus Diol (OH) OBD Preparative Column provides additional selectivity for chromatographic separations. Its high-density diol surface bonding delivers robust chromatography performance, comparable to traditional unbonded silica phases, especially when used with additives.

  • Features DIOL chemistry for versatile separations.
  • Exhibits good peak shapes for acidic compounds.
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles.
  • Offers a 130 Å pore size for optimal separation efficiency.
  • Utilizes a 5 μm particle size for high resolution.
  • Designed with 19 x 100 mm dimensions for preparative scale applications.

Catalog No. 50-239-2712


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.