Learn More
Medchemexpress LLC Tos-Gly-Pro-Arg-ANBA-IPA acetate | 2070009-46-8 | 99.2% | 747.82 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0020A10MG
Encompass_Preferred
Tos-Gly-Pro-Arg-ANBA-IPA acetate is a chromogenic peptide substrate designed for luminescence measurement. This product is intended for research use only and not sold to patients.
  • Chromogenic peptide substrate
  • Used for luminescence measurement
  • Intended for research use only
  • Not sold to patients
  • Appearance: solid
  • Color: off-white to light yellow
Catalog No. 50-004-26608