Learn More
Medchemexpress LLC Tos-Gly-Pro-Arg-ANBA-IPA (acetate) | 2070009-46-8 | 99.2% | 747.82 | 50MG
SDP
Supplier:  Medchemexpress LLC HYP0020A50MG
Encompass_Preferred
Tos-Gly-Pro-Arg-ANBA-IPA acetate is a chromogenic peptide substrate intended for research use only. Also known as tos-GPR-ANBA-IPA acetate, this solid compound appears off-white to light yellow and can be utilized for luminescence measurement. It is stable under recommended storage conditions, making it suitable for various laboratory applications.
  • Chromogenic peptide substrate
  • Suitable for luminescence measurement
  • Solid, off-white to light yellow appearance
  • Intended for research use only
  • Stable under recommended storage conditions
Catalog No. 50-004-25031