Learn More
Medchemexpress LLC Tos-PEG2-Boc | 1309666-78-1 | 98.0% | 1 G

Supplier: Medchemexpress LLC HY1305341G
Tos-PEG2-Boc is a PEG-based PROTAC linker, identified by CAS number 1309666-78-1, with a purity of 98.0%. This chemical is stable under recommended storage conditions and is intended for laboratory research use only.
- Functions as a PEG-based PROTAC linker.
- Presents as a white to off-white solid.
- Stable when stored under recommended conditions.
- Acceptable for shipping at room temperature for less than two weeks.
- Store powder at -20°C for three years or 4°C for two years.
- Store in solvent at -80°C for six months or -20°C for one month.
- Not fully validated for medical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.