Learn More
Medchemexpress LLC TOS-PEG2-O-PROPARGYL - 100MG
SDP
Supplier:  Medchemexpress LLC HY130162100MG
Encompass_Preferred
Tos-PEG2-O-Propargyl is a PEG-based PROTAC linker can be used in the synthesis of Thalidomide-O-PEG2-propargyl (HY-126458).Purity: 98.47%.CAS No. : 1119249-30-7
Catalog No. NC1904254