Learn More
Medchemexpress LLC Tos-PEG4-acid | 1453117-42-4 | 97.2% | 25 MG

Supplier: Medchemexpress LLC 25MGHY130417
Tos-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These PROTACs consist of two ligands linked together, one for an E3 ubiquitin ligase and the other for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. It has a molecular weight of 376.42 and a chemical formula of C16H24O8S, presenting as a colorless to light yellow liquid.
- PEG-based PROTAC linker
- Facilitates synthesis of PROTACs
- Molecular weight of 376.42
- Chemical formula: C16H24O8S
- Appears as a colorless to light yellow liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.