Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tos-PEG4-NH-Boc (PROTAC linker 7) | 1246999-33-6 | ≥98.0% | 10 G
SDP

Supplier:  Medchemexpress LLC HY12880310G

Encompass_Preferred

Tos-PEG4-NH-Boc (PROTAC Linker 7) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This chemical is intended for research use only.

  • Cas number: 1246999-33-6
  • Molecular weight: 447.54
  • Formula: C20H33NO8S
  • Purity: ≥98.0%

Catalog No. 50-003-40545


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.