Learn More
Medchemexpress LLC Tos-PEG4-NH-Boc (PROTAC Linker 7) | 1246999-33-6 | >98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128803100MG
Encompass_Preferred
Tos-PEG4-NH-Boc (PROTAC Linker 7) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker.
  • Used in the synthesis of PROTACs.
  • Connects two different ligands.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-003-41824