Learn More
Medchemexpress LLC Tos-PEG4-NH-Boc (PROTAC linker 7) | 1246999-33-6 | ≥98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1288035G
Encompass_Preferred
Tos-PEG4-NH-Boc (PROTAC Linker 7) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only. PROTACs are compounds with two ligands joined by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. They leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Intended for research use only
  • Targets an E3 ubiquitin ligase and a specific protein
  • Leverages the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-42919