Learn More
Medchemexpress LLC Tos-PEG4-THP | 86259-89-4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130819100MG
Encompass_Preferred
Tos-PEG4-THP is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTAC K-Ras Degrader-1. PROTACs are compounds containing two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. These compounds leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Composed of two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-38474