Learn More
Medchemexpress LLC Tos-Pro-OH | 51077-01-1 | MFCD00238392 | 99.0% | 269.32 g/mol | C12H15NO4S | 5 G

Supplier: Medchemexpress LLC HYW1420715G
Tos-Pro-OH is a tosyl-proline derivative (N-toluenesulfonyl-L-proline) supplied as a solid research reagent. It functions as a protected proline building block for organic and peptide synthesis and is identified by molecular formula C12H15NO4S, molecular weight 269.32 g/mol, and CAS 51077-01-1.
- Purity 99.0% for consistent performance.
- Tosyl-protected proline useful as a building block in organic and peptide synthesis.
- Solid form simplifies weighing and storage.
- Molecular formula C12H15NO4S and molecular weight 269.32 g/mol for unambiguous identification.
- Available in lab-scale package sizes suitable for synthesis workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.