Learn More
Cayman Chemical Tosyl Phenylalnyl CLOrometh
SDP
Supplier:  Cayman Chemical 20166500 MG
Encompass_Preferred
An irreversible inhibitor of chymotrypsin-like proteases that has been shown to affect cell proliferation, apoptosis, and tumorigenesis; disrupts PDK1 signaling to the AGC kinases and also inhibits superoxide production
Catalog No. 50-298-5021