Learn More
Medchemexpress LLC Tosylate-DPA-714 | 958233-17-5 | 97.0% | 10 MG

Supplier: Medchemexpress LLC HY40088210MG
Tosylate-DPA-714 is the tosylate analog of DPA-714 used as a precursor for synthesizing 18F-labeled DPA-714, a translocator protein (TSPO)-selective ligand for neuroinflammation imaging research. Supplied for research use only in small laboratory quantities.
- Precursor for 18F radiolabeling of DPA-714.
- High purity (HPLC), approximately 97.0%.
- Molecular formula C29H34N4O5S; molecular weight 550.67 g·mol⁻¹.
- Supplied in small quantities suitable for synthesis (e.g., 10 MG).
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.