Learn More
Medchemexpress LLC Tosylchloramide sodium trihydrate | 7080-50-4 | 281.69 | 100 G
SDP
Supplier:  Medchemexpress LLC HYU00087100G
Encompass_Preferred
Tosylchloramide sodium trihydrate (Chloramine T sodium trihydrate) is a disinfectant agent widely used in laboratories, kitchens, and hospitals. It is also utilized as a biocide in air fresheners and deodorants. This product is intended for research use only and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities.
  • Disinfectant agent
  • Used in laboratories, kitchens, and hospitals
  • Biocide in air fresheners and deodorants
  • Stable under recommended storage conditions
  • Identified as a laboratory chemical
  • Used in the manufacture of substances
  • Categorized as an inhibitor
  • Component of screening libraries
  • Appears as a solid
Catalog No. 50-004-49004