Learn More
Medchemexpress LLC Tosylchloramide sodium trihydrate | 7080-50-4 | 281.69 | 50 G

Supplier: Medchemexpress LLC HYU0008750G
Tosylchloramide sodium trihydrate (Chloramine T sodium trihydrate) is a disinfectant agent widely used in laboratories, kitchens, and hospitals. It is also utilized as a biocide in air fresheners and deodorants.
- Disinfectant agent for various settings
- Utilized as a biocide in air fresheners and deodorants
- Purity of 98.0%
- Molecular weight of 281.69
- Appearance as a white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.