Learn More
Cell Biolabs Inc OxiSelect™ Total Antioxidant Capacity (TAC) Assay Kit, 200 assays
Supplier:  Cell Biolabs Inc STA360
Encompass_Preferred
Cell Biolabs’ OxiSelect™ Total Antioxidant Capacity (TAC) Assay Kit measures the total antioxidant capacity of biomolecules from a variety of samples via a SET mechanism. The TAC Assay is based on the reduction of copper (II) to copper (I) by antioxidants such as uric acid. Upon reduction, the copper (I) ion further reacts with a coupling chromogenic reagent that produces a color with a maximum absorbance at 490 nm. Copper is advantageous over iron-based antioxidant assays because all classes of antioxidants, including thiols, are detected with marginal radical interference. Cell Biolabs’ OxiSelect™ TAC Assay Kit is a fast and reliable kit for the direct measurement of total antioxidant capacity from cell lysate, plasma, serum, urine, tissue homogenates, and food extracts. Each kit provides sufficient reagents to perform up to 200 assays, including blanks, antioxidant standards and unknown samples. Both hydrophilic and lipophilic samples are compatible with the assay.
Catalog No. NC0067485