Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys Total Bile Acids FS is a ready-to-use two component photometric reagent to determine Total Bile Acids in serum or heparin plasma on photometric systems. Each set contains R1= 46mL, R2=17mL
Supplier:  DIASYS DIAGNOSTIC SYSTEMS USA LLC 122389910921
Encompass_Preferred
Diagnostic reagent for quantitative in vitro determination of Total Bile Acids in animal serum, heparin plasma or stool samples on photometric systems. For veterinary or research use only. Serum TBA levels are a sensitive marker of liver function, reflecting hepatic synthesis, secretion and re-absorption. Compared to conventional liver screening tests such as ALT or AST, which indicate acute liver damage, the determination of total bile acids allows early detection of liver dysfunction and early treatment and prevention of severe, irreversible liver damage. Once a patient suffers from a liver disease, serum TBA can be used to monitor the treatment response. Although TBA levels provide early diagnosis of hepatobiliary deficiencies, they do not allow the differentiation between various diseases. Method: Enzymatic cycling method. Reagent 1 contains Buffer, Thio-NAD, reagent 2 contains Buffer, and 3-α-HSD. Reagents must be stored refrigerated.
Catalog No. NC2874342