Learn More
Enzo Life Sciences CYTAG TotalCGH Labeling kit (100Reactions)
SDP
Supplier:  Enzo Life Sciences ENZ426740100
Encompass_Preferred
Comparative genomic hybridization (aCGH) is a powerful diagnostic tool for detecting DNA copy number gains and losses associated with chromosome abnormalities. aCGH provides an understanding of genetic disorders, cancers and other genomic aberrations.   With optimized, proprietary reagents, Enzo's CYTAG CGH Labeling Kits produce high quality data using as little as 0.25µg of genomic DNA, without a need for pre-amplification. Bulk quantity kit, 2 x 1000 reactions, is available upon special request. Applications: CGH.
Catalog No. 50-201-2534
Add to Cart
Request a Quote