Learn More
Crystal Chem Inc Total Prostate-Specific Antigen (tPSA) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80976
Encompass
The Total Prostate-Specific Antigen (tPSA) ELISA is a highly sensitive assay that is used to quantify levels of human total PSA in serum. Highlights: Kit uses sample size of only 25 µL. High sensitivity of 0.1 ng/mL. Dynamic range of 0.5 - 100 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7878